Key Documents
Z286206
Michel-Miller HPLPLC starter kit
Analytical kit
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
manufacturer/tradename
Ace Glass 5795202
General description
The Michel-Miller chromatography system is designed for high-performance, low pressure liquid chromatography (HPLPLC) in analytical or preparative applications. These systems are for use at low (0-100psi) and moderate (100-300psi) pressures.
  • Constructed of epoxy-coated borosilicate glass and PTFE, without O-rings, leak free
  • Safe to 300psi, with safety shield
  • Usable with various packing materials
  • Zero dead volume
Convenient kits contain the basic items needed for operation. Other items such as pressure gauge, pump, valves, etc. are not included.
Kit Components Only
Product No.
Description
  • Safety shields, plastic, size A 2
  • Caps for size A safety shields 2
  • Michel-Miller column coupling, Ace-Thred for #7-11 1
  • Injection-port adapter, Complete, Ace-Thred for #7 1
  • TFE tubing for 1.5 mm i.d. tubing 3 m
  • TFE tubing for 2.0 mm i.d. tubing 3 m
Kit Components Also Available Separately
Product No.
Description
SDS
  • Z179477Michel-Miller chromatography column, L × I.D. 250 mm × 8 mm 1SDS
  • Z179485Michel-Miller chromatography column, L × I.D. 300 mm × 21 mm 1SDS
  • Z179620Michel-Miller end-fitting adapter, Ace-Thred for #7 2SDS
  • Z179639Bottom end fitting for Michel-Miller adjustable bed height column 2SDS
  • Z179981Michel-Miller column filter disc, Ace-Thred for #7 100SDS
  • Z180017Michel-Miller column filter disc, Ace-Thred for #11 100SDS
  • Z179965Stainless steel tubing adapter 2SDS
  • Tubing connector for large columns, Nut, tube I.D. 2 mm 12
  • Tubing connector for large columns, Insert, tube I.D. 2 mm 12
  • Z180351Tubing connector for large columns, Tubing insertion tool, tube I.D. 1.5 mm 1SDS
See All (10)
Choose from one of the most recent versions:
Certificates of Analysis (COA)