All Photos(1)
About This Item
Linear Formula:
21Ne
CAS Number:
Molecular Weight:
20.99
MDL number:
UNSPSC Code:
12142006
PubChem Substance ID:
Recommended Products
isotopic purity
90 atom %
Assay
≥99.5% (Total Neon)
form
gas
impurities
<0.5% Total others
mass shift
M+1
SMILES string
[21Ne]
InChI
1S/Ne/i1+1
InChI key
GKAOGPIIYCISHV-OUBTZVSYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service