Stem Cell Culture
Stem cells have the unique ability to self-renew or differentiate into various cell types in response to appropriate signals. These properties provide stem cells with unique capabilities for tissue repair, replacement and regeneration, making them valuable research tools in regenerative medicine and stem cell therapies.
Featured Categories
Discover our collection of validated human primary cells from PromoCell® human primary cells, primary blood and immune cells from BioIVT, and more.
Rich in growth-promoting factors, Fetal bovine serum (FBS) or fetal calf serum (FCS) is an ideal supplement for mammalian cell culture.
Seamless Sterile Filtration: Choose us for reliable Stericap® & Steripak® devices, ensuring sterile, quality solutions from mL to 20 L.
Next-gen Stericup® Quick Release: upgraded design enhances usability, streamlines filtration, safeguards cell cultures with Millipore® membranes.
Types and Features of Stem Cells
- Stem cells have unlimited self-renewal capabilities
- Stem cells are non-differentiated cells with unspecialized functions
- Stem cells can differentiate into specific cell types under the right conditions. Stem cells are broadly characterized into multipotent or pluripotent stem cells.
Multipotent stem cells include adult stem cells that can self-renew or to differentiate into specialized, tissue-specific cell types. Examples include hematopoietic stem cells (HSCs) that differentiate into various blood cells; mesenchymal stem cells (MSCs) that differentiate into osteoblasts, myocytes, chondrocytes and adipocytes; and neural stem cells (NSCs) that differentiate into neurons, astrocytes and oligodendrocytes.
Pluripotent stem cells can differentiate into any cell lineage. They are classified based on the tissue of origin into embryonic stem cells (ESCs), perinatal stem cells, and induced pluripotent stem cells (iPSCs). ESCs are derived from embryos and can divide indefinitely in an in vivo stem cell culture. Perinatal stem cells are derived from umbilical or placental blood or tissue, and are the most widely used pluripotent stem cells. Cord blood banking at birth is increasingly accepted as an option for treating complicated disorders later in life. The iPSCs are adult cells that are reprogrammed, or induced to behave like ESCs. The significant advantage of using iPSCs for medical applications is the reduced chance of graft rejection, since the cells are derived from the patient’s own tissues.
Stem Cell Research Applications
Stem cells occupy an active and growing area of basic science and clinical research due to their ability to self-renew and differentiate into mature cell types. Current clinical applications for stem cells include treatments for neurological and cardiovascular diseases, autoimmune disorders, cancer, wound healing, and disease modeling and drug screening. Newly discovered gene editing technologies like CRISPR may advance stem cell research and offer enormous promise in treating difficult disorders.
Stem Cell Culture Basics
Stem cells require specialized, high-quality media and expert culture techniques for propagation in the laboratory. Suboptimal stem cell culture conditions can easily lead to unwanted stem cell differentiation or to cellular senescence. Stem cell differentiation is triggered by various factors in vivo, some of which can be replicated in in vitro stem cell cultures. Some stem cell lines are immortal and can be cultured indefinitely, so it is imperative to select the right stem cell type for your research application.
Recent advances in the stem cell field have been due to the advent of CRISPR genome editing technology and 3D cell culture techniques. Advanced protocols such as those that generate organoids from iPSCs have provided scientists with more predictive in vitro “disease-in-a-dish” models.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- FAQs and information about IPS Cells or induced pluripotent stem cells (iPSCs). Including what are IPS cells, learning stem cell basics, reprogramming methods, and iPS cell culture techniques.
- Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).
- We offer answers to frequently asked questions and a large collection of cell culture media, supplements, and bioactive small molecules used to control the cell fate of human iPSCs.
- Read types of stem cells including multipotent stem cells, Pluripotent stem cells and iPSCs and their applications in basic stem cell research, stem cell therapy and disease modelling
- This white paper illustrates the importance of three-dimensional (3D) cell culture in the production of pluripotent stem cell (PSC)-derived models of embryo development and differentiation. It contains a general overview of embryoid bodies (EBs) and a comparison between the various methods used for their generation.
- See All (28)
Related Protocols
- Information about mesenchyme, specifically mesenchymal stem cell procotols. Step-by-step cell culture protocols for mesenchymal stem cell (MSC) isolation, expansion and differentiation.
- Step-by-step stem cell culture protocols for human induced pluripotent stem cells (iPSCs) including ips cell thawing, expanding, freezing and characterizing.
- Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.
- Stem Cell protocols for cryopreservation, thawing of cryopreserved stem cells and media preparation.
- Hematopoietic stem cell culture protocols for isolation, expansion, and differentiation of CD34+ cells including CFU assays.
- See All (9)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email [email protected]
or call +1 (800) 244-1173
Additional Support
- Chromatogram Search
Use the Chromatogram Search to identify unknown compounds in your sample.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ