HPLC Analysis of 3-Methylpyrazole and 4-Methylpyrazole in Urine on Discovery® C18 after SPE using Discovery® DSC-SCX
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
urine spiked with 3- and 4-methylpyrazole at 1 mg/mL
SPE tube/cartridge
Discovery DSC-SCX, 500 mg/3 mL (52686-U)
condition
2 mL methanol; 2 mL DI water
sample addition
1 mL
washing
2 mL DI water
elution
2 mL 5% methanol in 250 mM potassium phosphate buffer, pH 7.4
column
Discovery C18, 15 cm × 4.6 mm, 5 μm preceded by a 2 cm guard column and 0.5 μm frit filter (504955)
mobile phase
MeOH:5 mM phosphate buffer, pH 6 (20:80)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 220 nm
injection
25 μL, dilute urine extract
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany