Merck
  • A randomized controlled study on the efficacy of a novel combination vaccine against enzootic pneumonia (Mycoplasma hyopneumoniae) and porcine Circovirus type 2 (PCV2) in the presence of strong maternally derived PCV2 immunity in pigs.
A randomized controlled study on the efficacy of a novel combination vaccine against enzootic pneumonia (Mycoplasma hyopneumoniae) and porcine Circovirus type 2 (PCV2) in the presence of strong maternally derived PCV2 immunity in pigs.
BMC veterinary research (2017-04-09)
Panagiotis D Tassis, Ioannis Tsakmakidis, Vassileios G Papatsiros, Dimitrios Koulialis, Tom Nell, Georgia Brellou, Eleni D Tzika
ABSTRACT
Mycoplasma hyopneumoniae (M. hyo) and Porcine Circovirus Type 2 (PCV2) are major pathogens that cause significant health problems in swine worldwide. Maternal derived immunity (MDI) has been suggested as a significant immediate defence factor for newborn piglets and may interfere with piglet's vaccination-induced immunity. The study aimed to assess the efficacy of a novel combination vaccine (consisting of PCV2 subunits and inactivated M. hyo strain J), against PCV2 and M. hyo natural infection [Porcilis Vaccination significantly reduced the severity of lung lesions at slaughter (lesions of cranio-ventral pulmonary consolidation) (P < 0.001). The overall mean lung lesion score (LLS) was 9.6 in the vaccinated group and 12.2 in controls. The level of PCV2 viraemia was significantly reduced in vaccinated pigs. Furthermore, 25 g higher average daily weight gain (ADWG) was observed during the finishing phase (P < 0.001) and 18 g greater ADWG overall (P < 0.001). Results of LLS, PCV2 viremia and ADWG support the test product's efficacy in the face of strong maternally derived PCV2 immunity.
Sigma-Aldrich
Human APP / Amyloid Beta A4 Protein ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.