Merck
  • Determination of linear and cyclic volatile methylsiloxanes in blood of turtles, cormorants, and seals from Canada.
Determination of linear and cyclic volatile methylsiloxanes in blood of turtles, cormorants, and seals from Canada.
The Science of the total environment (2016-09-25)
De-Gao Wang, Shane R de Solla, Michel Lebeuf, Tommy Bisbicos, Glenn C Barrett, Mehran Alaee
ABSTRACT
We measured the concentrations of linear and cyclic volatile methylsiloxanes (VMS) concentrations in the blood plasma of turtles, cormorants, and seals collected from Canadian freshwater and marine ecosystems. A modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) method was developed to quantify the levels of linear and cyclic VMS in the plasma samples. The cyclic VMS of hexamethylcyclotrisiloxane (D3), octamethylcyclotetrasiloxane (D4), decamethylcyclopentasiloxane (D5), and dodecamethylcyclohexasiloxane (D6) were present in the plasma of all three species. Linear VMS compounds were observed only in seal plasma from one contaminated site. There was no statistically significant difference among species and locations for D3, D4, and D6 concentrations. Average D5 concentrations ranged from 0.143 to 7.39ngg
Sigma-Aldrich
Tetrakis(trimethylsilyloxy)silane, 97%
Sigma-Aldrich
Decamethyltetrasiloxane, 97%
Sigma-Aldrich
Dodecamethylpentasiloxane, 97%