Merck
Photosynthetic leaf area modulates tiller bud outgrowth in sorghum.
Plant, cell & environment (2014-12-17)
Tesfamichael H Kebrom, John E Mullet
ABSTRACT
Shoot branches or tillers develop from axillary buds. The dormancy versus outgrowth fates of buds depends on genetic, environmental and hormonal signals. Defoliation inhibits bud outgrowth indicating the role of leaf-derived metabolic factors such as sucrose in bud outgrowth. In this study, the sensitivity of bud outgrowth to selective defoliation was investigated. At 6 d after planting (6 DAP), the first two leaves of sorghum were fully expanded and the third was partially emerged. Therefore, the leaves were selectively defoliated at 6 DAP and the length of the bud in the first leaf axil was measured at 8 DAP. Bud outgrowth was inhibited by defoliation of only 2 cm from the tip of the second leaf blade. The expression of dormancy and sucrose-starvation marker genes was up-regulated and cell cycle and sucrose-inducible genes was down-regulated during the first 24 h post-defoliation of the second leaf. At 48 h, the expression of these genes was similar to controls as the defoliated plant recovers. Our results demonstrate that small changes in photosynthetic leaf area affect the propensity of tiller buds for outgrowth. Therefore, variation in leaf area and photosynthetic activity should be included when integrating sucrose into models of shoot branching.
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, iQ, with fluorescein for Bio-Rad systems
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, Low ROX, for ABI and Stratagene instruments
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, For Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
tert-Butanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
tert-Butanol, SAJ first grade, ≥98.0%