- Ionic liquids based on N-alkyl-N-methylmorpholinium salts as potential electrolytes.
Ionic liquids based on N-alkyl-N-methylmorpholinium salts as potential electrolytes.
Chemical communications (Cambridge, England) (2004-03-27)
Ki-Sub Kim, Sukjeong Choi, D Demberelnyamba, Huen Lee, Jaeseung Oh, Byoung-Bae Lee, Su-Jin Mun
PMID15045084
ABSTRACT
Ionic liquids based on N-alkyl-N-methylmorpholinium salts have been synthesized and the physical and electrochemical characteristics of this family of ionic liquids have been investigated for use as electrolytes.