Merck
  • [Distribution of 14C-phenazepam and its metabolites in the liver, brain and blood plasma in single and prolonged administration to white rats].
[Distribution of 14C-phenazepam and its metabolites in the liver, brain and blood plasma in single and prolonged administration to white rats].
Farmakologiia i toksikologiia (1980-03-01)
N Ia Golovenko, V G Zin'kovskiĭ, S B Seredenin, S A Andronati, L N Iakubovskaia
ABSTRACT
The distribution pattern of 14C-phenazepam and some of its metabolites in the liver, brain and blood plasma at a prolonged (45 days) administration to rats does not differ from that after a single administration. The liver and brain demonstrated an increase in the content of 3-hydroxyderivative. Glycuronide was found only in the liver.
Product Description
Supelco
Phenazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Phenazepam, ≥97% (HPLC)