Merck
  • Adaptation of innate lymphoid cells to a micronutrient deficiency promotes type 2 barrier immunity.
Adaptation of innate lymphoid cells to a micronutrient deficiency promotes type 2 barrier immunity.
Science (New York, N.Y.) (2014-01-25)
S P Spencer, C Wilhelm, Q Yang, J A Hall, N Bouladoux, A Boyd, T B Nutman, J F Urban, J Wang, T R Ramalingam, A Bhandoola, T A Wynn, Y Belkaid
ABSTRACT
How the immune system adapts to malnutrition to sustain immunity at barrier surfaces, such as the intestine, remains unclear. Vitamin A deficiency is one of the most common micronutrient deficiencies and is associated with profound defects in adaptive immunity. Here, we found that type 3 innate lymphoid cells (ILC3s) are severely diminished in vitamin A-deficient settings, which results in compromised immunity to acute bacterial infection. However, vitamin A deprivation paradoxically resulted in dramatic expansion of interleukin-13 (IL-13)-producing ILC2s and resistance to nematode infection in mice, which revealed that ILCs are primary sensors of dietary stress. Further, these data indicate that, during malnutrition, a switch to innate type 2 immunity may represent a powerful adaptation of the immune system to promote host survival in the face of ongoing barrier challenges.
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg