Merck
  • Synthesis and biological activity of conformationally restricted analogs of milnacipran: (1S,2R)-1-phenyl-2-[(S)-1-aminopropyl]-N,N-diethylcyclopropanecarboxami de, an efficient noncompetitive N-methyl-D-aspartic acid receptor antagonist.
Synthesis and biological activity of conformationally restricted analogs of milnacipran: (1S,2R)-1-phenyl-2-[(S)-1-aminopropyl]-N,N-diethylcyclopropanecarboxami de, an efficient noncompetitive N-methyl-D-aspartic acid receptor antagonist.
Journal of medicinal chemistry (1996-11-22)
S Shuto, S Ono, Y Hase, Y Ueno, T Noguchi, K Yoshii, A Matsuda
ABSTRACT
We recently demonstrated that (+/-)-(Z)-2-(aminomethyl)-1-phenyl-N,N-diethylcyclopropanecarboxamide [milnacipran, (+/-)-1], an inhibitor of the reuptake of serotonin (5-HT), was a noncompetitive NMDA receptor antagonist. On the basis of the cyclopropane structure of (+/-)-1, conformationally restricted analogs with different stereochemistries, namely 1-phenyl-2-(1-aminoalkyl)-N,N-diethylcyclopropanecarboxamindes (2, 3, ent-2, and ent-3), were designed and synthesized. Among these analogs, 2a, 2b, and 2f, with (1S,2R,1'S)-configuration, were more efficient than milnacipran as NMDA receptor antagonists; these compounds significantly inhibited the binding of [3H]MK-801 at IC50 = 0.35 +/- 0.08, 0.20 +/- 0.024, and 0.16 +/- 0.02 microM, respectively, and blocked the response of voltage-clamped oocytes to NMDA, surpassing the effects of (+/-)-1. Although both the 1'-methyl analog 2a and the 1'-vinyl analog 2f, like (+/-)-1, strongly inhibited 5-HT uptake in vitro, the corresponding 1'-ethyl analog 2b was devoid of the inhibitory effect on 5-HT uptake, while it was about 30 times more potent as an NMDA receptor antagonist than (+/-)-1.
Sigma-Aldrich
Cyclopropanecarboxylic acid, 95%