Merck
  • Trisaccharide synthesis by glycosyl transfer from p-nitrophenyl beta-D-N-acetylgalactosaminide on to disaccharide acceptors catalysed by the beta-N-acetylhexosaminidase from Aspergillus oryzae.
Trisaccharide synthesis by glycosyl transfer from p-nitrophenyl beta-D-N-acetylgalactosaminide on to disaccharide acceptors catalysed by the beta-N-acetylhexosaminidase from Aspergillus oryzae.
Carbohydrate research (1998-07-02)
S Singh, M Scigelova, P Critchley, D H Crout
ABSTRACT
The beta-N-acetylhexosaminidase from Aspergillus oryzae catalysed the transfer of beta-D-N-acetylgalactosaminyl residues from p-nitrophenyl beta-D-N-acetylglucosaminide on to disaccharide acceptors consisting of thioethyl glycosides of alpha-D-Glc-(1-->4)-beta-D-Glc, beta-D-Glc-(1-->4)-beta-D-Glc and beta-D-Glc-(1-->6)-beta-D-Glc. The principle of 'anomeric control' was exemplified by the results which showed that an alpha-linkage between the units of the acceptor favoured exclusively the formation of a new (1-->4)-linkage, whereas the beta-configuration in the acceptor led to a mixture of (1-->4)- and (1-->3)-linked products, as observed for simple glycosides of monosaccharide acceptors. With the thioethyl beta-lactoside as acceptor, beta-D-Gal-(1-->6)-beta-D-Gal-(1-->4)-beta-D-GlcSEt was formed, owing to the action of residual beta-D-galactosidase activity in the N-acetylhexosaminidase on the thioethyl beta-lactoside acting as both donor and acceptor.
Sigma-Aldrich
4-Nitrophenyl N-acetyl-α-D-galactosaminide, substrate for N-acetyl-α-D-galactosaminidase
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-galactosaminide, ≥98%