Merck
  • Tuning the functionality of a carbon nanofiber-Pt-RuO2 system from charge storage to electrocatalysis.
Tuning the functionality of a carbon nanofiber-Pt-RuO2 system from charge storage to electrocatalysis.
Inorganic chemistry (2012-09-06)
Beena K Balan, Sreekumar Kurungot
ABSTRACT
Chemical-functionalization-induced switching in the property of a hybrid system composed of a hollow carbon nanofiber (CNF) and Pt and RuO(2) nanoparticles from charge storage to electrocatalysis is presented. The results of this study show how important it is to have a clear understanding of the nature of surface functionalities in the processes involving dispersion of more than one component on various substrates including carbon nanomorphologies. When pristine CNF is used to decorate Pt and RuO(2) nanoparticles, random dispersion occurs on the CNF surface (C-PtRuO(2)). This results in mainly phase-separated nanoparticles rich in RuO(2) characteristics. In contrast to this, upon moving from the pristine CNF to those activated by a simple H(2)O(2) treatment to create oxygen-containing surface functional groups, a material rich in Pt features on the surface is obtained (F-PtRuO(2)). This is achieved because of the preferential adsorption of RuO(2) by the functionalized surface of CNF. A better affinity of the oxygen-containing functional groups on CNF toward RuO(2) mobilizes relatively faster adsorption of this moiety, leading to a well-controlled segregation of Pt nanoparticles toward the surface. Further reorganization of Pt nanoparticles leads to the formation of a Pt nanosheet structure on the surface. The electrochemical properties of these materials are initially evaluated using cyclic voltammetric analysis. The cyclic voltammetric results indicate that C-PtRuO(2) shows a charge storage property, a typical characteristic of hydrous RuO(2), whereas F-PtRuO(2) shows an oxygen reduction property, which is the characteristic feature of Pt. This clear switch in the behavior from charge storage to electrocatalysis is further confirmed by galvanostatic charge-discharge and rotating-disk-electrode studies.
Sigma-Aldrich
Ruthenium(IV) oxide hydrate, powder
Sigma-Aldrich
Ruthenium(IV) oxide, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium(IV) oxide hydrate, 99.9% trace metals basis