Merck
  • Coated chitosan pellets containing rutin intended for the treatment of inflammatory bowel disease: in vitro characteristics and in vivo evaluation.
Coated chitosan pellets containing rutin intended for the treatment of inflammatory bowel disease: in vitro characteristics and in vivo evaluation.
International journal of pharmaceutics (2011-11-15)
Miloslava Rabišková, Tereza Bautzová, Jan Gajdziok, Kateřina Dvořáčková, Alf Lamprecht, Yann Pellequer, Jiřina Spilková
ABSTRACT
Preparation of coated pellets intended for rutin colon delivery, their evaluation in vitro and in vivo in experimental colitis in rats was the purpose of this study. Pellets were obtained using extrusion/spheronization and coated with three types of coatings (caffeic acid/hypromellose/alginic acid; sodium alginate/hypromellose/zinc acetate; sodium alginate/chitosan). Dissolution using buffers of pH values, β-glucosidase and times corresponding to gastrointestinal tract (GIT) was provided. Pellets coated with alginate/chitosan showed low rutin dissolution (12-14%) in upper GIT conditions and fast release (87-89%) under colon conditions; that is a good presumption of intended rutin release. After colitis induction and development, the rats were treated with pellets and rutin solution administered orally, solution also rectally. Colon/body weight ratio, myeloperoxidase activity and histological evaluation were performed. Rutin was able to promote colonic healing at the dose of 10mg/kg: colon/body weight ratio decreased and myeloperoxidase activity was significantly suppressed. Pellets coated with alginate/chitosan applied orally and rutin solution administered rectally showed the best efficacy. The combination of rutin as natural product, mucoadhesive chitosan degraded in the colon and sodium alginate as the main coating substance in the form of pellets create a promising preparation for therapy of this severe illness.
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)