Merck
  • Protein immobilisation on perpendicularly aligned gold tipped nanorod assemblies.
Protein immobilisation on perpendicularly aligned gold tipped nanorod assemblies.
Chemical communications (Cambridge, England) (2011-01-12)
Catriona O'Sullivan, Suzanne Crilly, Fathima R Laffir, Ajay Singh, Edmond Magner, Kevin M Ryan
ABSTRACT
A multi component assembly consisting of the redox protein cytochrome c (cyt c) immobilised onto vertically aligned gold tipped semiconductor nanorods is described. Cyt c was successfully immobilised using a thiol linker. A faradaic response demonstrated that the protein is electroactive in this ultra high density array.
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Cadmium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)