Merck
  • Phenolic compounds from Pueraria lobata protect PC12 cells against Aβ-induced toxicity.
Phenolic compounds from Pueraria lobata protect PC12 cells against Aβ-induced toxicity.
Archives of pharmacal research (2010-11-06)
Yun-hyeok Choi, Seong Su Hong, Yu Su Shin, Bang Yeon Hwang, So-Young Park, Dongho Lee
ABSTRACT
Bioassay-guided fractionation of the EtOAc-soluble extract of Pueraria lobata based on the inhibition of Aβ-induced toxicity in PC12 cells resulted in the isolation of four known active compounds, genistein (8), biochanin A (9), sissotrin (10), and puerol B (11). Of these, genistein (8) and biochanin A (9) exhibited potent neuroprotective effects with ED(50) values of 33.7 and 27.8 μM, respectively. In addition, a new coumestan, 2-(α,α-dimethylallyl)coumestrol (1) was isolated and characterized, but proved to be inactive, as were additional seven known compounds. The structure of new compound 1 was determined using spectroscopic techniques.
Sigma-Aldrich
Coumestrol, BioReagent, suitable for fluorescence, ≥97.5% (HPLC)
Sigma-Aldrich
Coumestrol, ≥95.0% (HPLC)