Merck
  • Classification of benzoyl peroxide as safe and effective and revision of labeling to drug facts format; topical acne drug products for over-the-counter human use; final rule.
Classification of benzoyl peroxide as safe and effective and revision of labeling to drug facts format; topical acne drug products for over-the-counter human use; final rule.
Federal register (2010-04-14)
ABSTRACT
We, the Food and Drug Administration (FDA), are issuing this final rule to include benzoyl peroxide as a generally recognized as safe and effective (GRASE) active ingredient in over-the-counter (OTC) topical acne drug products. In addition, this final rule includes new warnings and directions required for OTC acne drug products containing benzoyl peroxide. We are also revising labeling for OTC topical acne drug products containing resorcinol, resorcinol monoacetate, salicylic acid and/or sulfur to meet OTC drug labeling content and format requirements in a certain FDA regulation. This final rule is part of our ongoing review of OTC drug products and represents our conclusions on benzoyl peroxide in OTC acne drug products.
Sigma-Aldrich
Resorcinol monoacetate, technical grade
Pricing and availability is not currently available.