Merck
  • [Development of a laminar bioadsorbent with hypertolerant Phanerochaete chrysosporium to Cd, Ni and Pb for wastewater treatment].
[Development of a laminar bioadsorbent with hypertolerant Phanerochaete chrysosporium to Cd, Ni and Pb for wastewater treatment].
Revista iberoamericana de micologia (2010-03-30)
Diana Morales-Fonseca, Katherine Ruiz-Tovar, María Mercedes Martínez-Salgado, Ana Bertha Soto-Guzmán, Ciro Falcony-Guajardo, Refugio Rodríguez Vázquez, Aura Marina Pedroza-Rodríguez
ABSTRACT
The use of basidiomycetes for metal removal is an alternative to traditional methods. In this, the biomass acts as a natural ionic exchanger removing metals from solution. To develop a laminar biosorbent using a basidiomycete fungus resistant to high Cd, Ni and Pb concentrations. The tolerance of Trametes versicolor, Pleurotus ostreatus and Phanerochaete chrysosporium was evaluated using increasing concentrations of the heavy metal salts, cadmium sulphate, lead acetate and nickel chloride. A biosorbent system was developed based on polyethylene sheets with a fungal biomass. It was evaluated in bubble columns using synthetic wastewater with the 3 metal salts at a rate of 300 mg/l. Finally, in a complementary experiment using shake flasks, the effect of a higher amount of biomass related to the metal removal efficiency was evaluated. P. chrysosporium strain was more tolerant to C₄H₆O₄Pb (10,000mg/l), Cl₂Ni (300mg/l) and CdSO₄·8H₂O (1,500 mg/l). In a reactor, under non-ligninolytic conditions, the fungus removed 69% of the chemical oxygen demand and produced enzymes such as LiP (0.01 U/l) and MnP (0.6 U/l.). An accumulation of metals in the wall was observed. By increasing the biomass to 1.6 (w/v), the metal biosorption was favored in the mixture (57% Pb, 74% Cd, and 98% Ni) and separately (95% Pb, 60% Cd, and 56% Ni). Competition between Ni and Pb by ligands of the wall was observed. A novel laminar system based on P. chrysosporium viable biomass was developed. It has a large surface area and tolerance to high concentrations of Cd, Ni and Pb. It seems to be an alternative for the removal of metals from water.
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Cadmium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)