Merck
  • Configurational assignment of carbon, silicon and germanium containing propynal oximes by means of 13C-1H, 13C-13C and 15N-1H spin-spin coupling constants.
Configurational assignment of carbon, silicon and germanium containing propynal oximes by means of 13C-1H, 13C-13C and 15N-1H spin-spin coupling constants.
Magnetic resonance in chemistry : MRC (2007-06-15)
Kirill A Chernyshev, Leonid B Krivdin, Lyudmila I Larina, Tatiana V Konkova, Maria M Demina, Alevtina S Medvedeva
ABSTRACT
Configurational assignment of five carbon, silicon and germanium containing propynal oximes has been carried out by means of experimental measurements and high-level ab initio calculations of their 13C-1H, 13C-13C and 15N-1H spin-spin coupling constants. The title compounds were shown to exist in the nonequilibrium mixture of E and Z isomers with the energy difference of less than 0.3 kcal/mol calculated at the MP2/6-311G** level.
Sigma-Aldrich
Propyne, ≥99%
Sigma-Aldrich
Propyne, ≥97%