Merck
  • Metabolism of carveol and dihydrocarveol in Rhodococcus erythropolis DCL14.
Metabolism of carveol and dihydrocarveol in Rhodococcus erythropolis DCL14.
Microbiology (Reading, England) (2000-06-01)
M J van der Werf, A M Boot
ABSTRACT
Rhodococcus erythropolis DCL14 assimilates all stereoisomers of carveol and dihydrocarveol as sole source of carbon and energy. Induction experiments with carveol- or dihydrocarveol-grown cells showed high oxygen consumption rates with these two compounds and with carvone and dihydrocarvone. (Dihydro)carveol-grown cells of R. erythropolis DCL14 contained the following enzymic activities involved in the carveol and dihydrocarveol degradation pathways of this micro-organism: (dihydro)carveol dehydrogenase (both NAD+- and dichlorophenolindophenol-dependent activities), an unknown cofactor-dependent carvone reductase, (iso-)dihydrocarvone isomerase activity, NADPH-dependent dihydrocarvone monooxygenase (Baeyer-Villiger monooxygenase), epsilon-lactone hydrolase and an NAD+-dependent 6-hydroxy-3-isopropenylheptanoate dehydrogenase. Product accumulation studies identified (4R)-carvone, (1R,4R)-dihydrocarvone, (4R,7R)-4-isopropenyl-7-methyl-2-oxo-oxepanone, (3R)-6-hydroxy-3-isopropenylheptanoate, (3R)-3-isopropenyl-6-oxoheptanoate, (3S,6R)-6-isopropenyl-3-methyl-2-oxooxepanone and (5R)-6-hydroxy-5-isopropenyl-2-methylhexanoate as intermediates in the (4R)-carveol degradation pathway. The opposite stereoisomers of these compounds were identified in the (4S)-carveol degradation pathway. With dihydrocarveol, the same intermediates are involved except that carvone was absent. These results show that R. erythropolis DCL14 metabolizes all four diastereomers of carveol via oxidation to carvone, which is subsequently stereospecifically reduced to (1R)-(iso-) dihydrocarvone. At this point also dihydrocarveol enters the pathway, and this compound is directly oxidized to (iso-)dihydrocarvone. Cell extracts contained both (1R)-(iso-)dihydrocarvone 1,2-monooxygenase and (1S)-(iso)-dihydrocarvone 2,3-monooxygenase activity, resulting in a branch point of the degradation pathway; (1R)-(iso-)dihydrocarvone was converted to 4-isopropenyl-7-methyl-2-oxo-oxepanone, while (1S)-(iso)-dihydrocarvone, which in vivo is isomerized to (1R)-(iso-)dihydrocarvone, was converted to 6-isopropenyl-3-methyl-2-oxo-oxepanone. 4-Isopropenyl-7-methyl-2-oxooxepanone is hydrolysed to 6-hydroxy-3-isopropenylheptanoate, which is subsequently oxidized to 3-isopropenyl-6-oxoheptanoate, thereby linking the (dihydro)carveol degradation pathways to the limonene degradation pathway of this micro-organism. 6-Isopropenyl-3-methyl-2-oxo-oxepanone is, in vitro, hydrolysed to 6-hydroxy-5-isopropenyl-2-methylhexanoate, which is thought to be a dead-end metabolite.
Sigma-Aldrich
L-Carveol, mixture of cis and trans, ≥95%, FG
Sigma-Aldrich
(−)-Carveol, mixture of isomers, 97%