Merck
  • Potassium channel Kir 4.1 regulates oligodendrocyte differentiation via intracellular pH regulation.
Potassium channel Kir 4.1 regulates oligodendrocyte differentiation via intracellular pH regulation.
Glia (2022-07-02)
Na Wang, Liang Zhou, Chong-Yu Shao, Xin-Tai Wang, Nan Zhang, Jiao Ma, Hai-Lan Hu, Yin Wang, Mengsheng Qiu, Ying Shen
ABSTRACT
In humans, loss-of-function mutations of Kcnj10 in SeSAME/EAST syndrome, which encodes the inwardly rectifying K+ channel 4.1 (Kir 4.1), causes progressive neurological decline. Despite its rich expression in oligodendrocyte (OL) lineage cells and an emerging link with demyelinating disease, the function of Kir 4.1 in OLs is unclear. Here we show a novel role of Kir 4.1 in OL development. Kir 4.1 expression is markedly greater in OLs than in OL precursor cells (OPCs), and the down-regulation of Kir 4.1 impairs OL maturation by affecting OPC differentiation. Interestingly, Kir 4.1 regulates the intracellular pH of OPCs and OLs via the Na+ /H+ exchanger, which underlies impeded OPC differentiation by Kir 4.1 inhibition. Furthermore, Kir 4.1 regulates GSK3β and SOX10, two molecules critical to OPC development. Collectively, our work opens a new avenue to understanding the functions of Kir 4.1 and intracellular pH in OLs.
Sigma-Aldrich
Anti-Sox10 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse