Merck
ER Stress Response in Human Cellular Models of Senescence.
The journals of gerontology. Series A, Biological sciences and medical sciences (2014-08-26)
Liliana Matos, Alexandra Monteiro Gouveia, Henrique Almeida
ABSTRACT
The aging process is characterized by progressive accumulation of damaged biomolecules in the endoplasmic reticulum, as result of increased oxidative stress accompanying cellular senescence. In agreement, we hypothesized that WI-38 human cellular models of replicative senescence and stress-induced premature senescence (SIPS) induced by hydrogen peroxide (H2O2-SIPS) or copper sulfate (CuSO4-SIPS) would present endoplasmic reticulum chaperoning mechanisms impairment and unfolded protein response activation. Results show that in replicative senescence and CuSO4-SIPS, immunoglobulin binding protein, calnexin, protein disulfide isomerase, and ER oxireductin-1 levels adjust to restore proteostasis and inositol-requiring enzyme-1 (IRE1)-, activating transcription factor 6 (ATF6)-, and pancreatic ER kinase (PERK)-mediated unfolded protein response are activated. However, H2O2-SIPS does not exhibit IRE1 and ATF6 pathways activation but a PERK-mediated upregulation of CCAAT/enhancer-binding protein homologous protein, showing that CuSO4-SIPS mimics better the endoplasmic reticulum molecular events of replicative senescence than H2O2-SIPS. Moreover, unfolded protein response activation is required for both SIPS models induction, because PERK and IRE1 inhibitors decreased senescence-associated beta-galactosidase appearance. In CuSO4-SIPS, the decrease in senescence levels is associated with PERK-driven, but IRE1 independent, cell cycle arrest while in H2O2-SIPS cell proliferation is PERK independent. These results add a step further on the molecular mechanisms that regulate senescence induction; moreover, they validate CuSO4-SIPS model as a useful tool to study cellular stress responses during aging, hoping to postpone age-related health decline.
Sigma-Aldrich
IRE1 Inhibitor III, 4μ8C, IRE1 Inhibitor III, CAS 14003-96-4, is a cell-permeable. Covalent inhibitor of IRE1 RNase activity (IC₅₀ = 550 and 45 nM, respectively, with 0 & 16 min preincubation in RNA cleavage assays).
Sigma-Aldrich
PERK Inhibitor I, GSK2606414, GSK2606414 is a cell-permeable, highly potent inhibitor of EIF2AK3/PERK (IC₅₀ = 0.4 nM; [ATP] = 5 µM). Targets PERK in its inactive DFG conformation at the ATP-binding region.