Merck
  • Electron transfer between cytochrome c and microsomal monooxygenase generates reactive oxygen species that accelerates apoptosis.
Electron transfer between cytochrome c and microsomal monooxygenase generates reactive oxygen species that accelerates apoptosis.
Redox biology (2022-05-25)
Han Xie, Li Song, Sagie Katz, Jinyu Zhu, Yawen Liu, Jinping Tang, Linjun Cai, Peter Hildebrandt, Xiao Xia Han
ABSTRACT
Generation of reactive oxygen species (ROS) are possibly induced by the crosstalk between mitochondria and endoplasmic reticula, which is physiologically important in apoptosis. Cytochrome c (Cyt c) is believed to play a crucial role in such signaling pathway by interrupting the coupling within microsomal monooxygenase (MMO). In this study, the correlation of ROS production with the electron transfer between Cyt c and the MMO system is investigated by resonance Raman (RR) spectroscopy. Binding of Cyt c to MMO is found to induce the production of ROS, which is quantitatively determined by the in-situ RR spectroscopy reflecting the interactions of Cyt c with generated ROS. The amount of ROS that is produced from isolated endoplasmic reticulum depends on the redox state of the Cyt c, indicating the important role of oxidized Cyt c in accelerating apoptosis. The role of electron transfer from MMO to Cyt c in the apoptotic mitochondria-endoplasmic reticulum pathway is accordingly proposed. This study is of significance for a deeper understanding of how Cyt c regulates apoptotic pathways through the endoplasmic reticulum, and thus may provide a rational basis for the design of antitumor drugs for cancer therapy.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Peroxide Assay Kit, sufficient for 250 colorimetric tests
Sigma-Aldrich
Endoplasmic Reticulum Isolation Kit, isolation of intact ER from mammalian soft tissues and cultured cells
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture