Merck
  • Toxic effects of dietary copper and EGCG on bioaccumulation, antioxidant enzyme and immune response of Korean bullhead, Pseudobagrus fulvidraco.
Toxic effects of dietary copper and EGCG on bioaccumulation, antioxidant enzyme and immune response of Korean bullhead, Pseudobagrus fulvidraco.
Fish & shellfish immunology (2021-01-28)
Huisu Lee, Jun-Hwan Kim, Hee-Ju Park, Ju-Chan Kang
ABSTRACT
There are few reports of dietary Cu (copper) toxicity to Korean bullhead, Pseudobagrus fulvidraco, and little is known about recovery from dietary Cu exposure. In this study, P. fulvidraco (mean length 16.9 ± 1.38 cm, and mean weight 53.2 ± 1.22 g) were exposed for 4 weeks to dietary Cu concentration of 0 (control), 700, 900, and 1100 mg Cu kg-1 dry feed to establish maximum tolerable levels of dietary Cu. All fish were then fed the dietary EGCG (Epigallocatechin gallate) concentration of 100 and 500 mg EGCG kg-1 dry feed for a further 2 weeks to assess recovery. We were measured bioaccumulation (in the intestine, liver, and gill tissue), antioxidant enzymes (SOD and CAT) and immune responses (lysozyme and phagocytosis). The Cu exposure induced a significant accumulation in the intestine, liver, and gill tissues and the highest accumulation was observed in intestinal tissues (17-34 fold), but dietary EGCG exposure decreased (about 0.8-fold) Cu concentration in each tissue (ANOVA, P < 0.05). In antioxidant enzymes, SOD and CAT significantly increased by approximately 1.6-fold by dietary Cu exposure in the liver and gill tissue, respectively, but dietary EGCG exposure decreased SOD and CAT by about 1.1-fold, respectively (ANOVA, P < 0.05). For immune responses, lysozyme and phagocytosis in the blood significantly were decreased by approximately 1.5-fold, respectively, by dietary Cu exposure, but dietary EGCG exposure increased lysozyme and phagocytosis by about 1.1-fold, respectively (ANOVA, P < 0.05). During recovery period, bioaccumulation, antioxidant enzymes (SOD and CAT activity), and immune response (lysozyme and phagocytosis activity) tended to alleviate the significant changes by Cu exposure, and the tendency to return normal state was observed in high level of EGCG. The result of this study indicate that Cu exposure to P. fulvidraco affects bioaccumulation, antioxidant enzymes, and immune responses, and high level of EGCG were effective to alleviate the toxic effects of Cu exposure.
Sigma-Aldrich
Cobalt(II) sulfate hydrate, 99.998% trace metals basis
Sigma-Aldrich
Calcium iodate, 98%