Merck
  • Generation of an iPSC line (AKOSi004-A) from fibroblasts of a female adult NPC1 patient, carrying the compound heterozygous mutation p.Val1023Serfs*15/p.Gly992Arg and of an iPSC line (AKOSi005-A) from a female adult control individual.
Generation of an iPSC line (AKOSi004-A) from fibroblasts of a female adult NPC1 patient, carrying the compound heterozygous mutation p.Val1023Serfs*15/p.Gly992Arg and of an iPSC line (AKOSi005-A) from a female adult control individual.
Stem cell research (2020-12-29)
Christin Völkner, Maik Liedtke, Janine Petters, Jan Lukas, Hugo Murua Escobar, Gudrun Knuebel, Jörn Bullerdiek, Carsten Holzmann, Andreas Hermann, Moritz J Frech
ABSTRACT
Niemann-Pick disease Type C (NPC) is a rare progressive neurodegenerative disorder with an incidence of 1:120,000 caused by mutations in the NPC1 or NPC2 gene leading to a massive cholesterol accumulation. Here, we describe the generation of induced pluripotent stem cells (iPSCs) of an affected female adult individual carrying the NPC1 mutation p.Val1023Serfs*15/p.Gly992Arg and an iPSC line from an unrelated healthy female adult control individual. Human iPSCs were derived from fibroblasts using retroviruses carrying the four reprogramming factors OCT4, SOX2, KLF4 and C-MYC. These lines provide a valuable resource for studying the pathophysiology of NPC and for pharmacological intervention.
Sigma-Aldrich
Ethenesulfonyl fluoride, 95%