Merck
  • Hormonal Regulation of Autophagy in Thyroid PCCL3 Cells and the Thyroids of Male Mice.
Hormonal Regulation of Autophagy in Thyroid PCCL3 Cells and the Thyroids of Male Mice.
Journal of the Endocrine Society (2020-07-17)
Tomomi Kurashige, Yasuyo Nakajima, Mika Shimamura, Masanobu Yamada, Yuji Nagayama
ABSTRACT
Autophagy is an evolutionarily conserved catabolic process by which cells degrade intracellular proteins and organelles in the lysosomes and recycle their metabolites. We have recently demonstrated the crucial role for the basal level of autophagic activity in thyrocyte survival and homeostasis using the thyroid-specific autophagy knockout mice. Here, we first studied hormonal regulation of autophagy in thyrocytes in vitro using a rat thyroid cell line PCCl3 and in vivo with mice. In cultured PCCl3 cells, thyroxine decreased microtubule-associated protein 1 light chain 3 (LC3) puncta (a component of autophagosome) and increased p62 (an autophagy substrate) levels, showing thyroxine-suppression of autophagy. In contrast, TSH increased both LC3 puncta and p62 levels, but at the same time stabilized p62 protein by inhibiting p62 degradation, indicating TSH induction of autophagy. Our experiments with various inhibitors identified that both the cAMP-protein kinase (PK) A-cAMP response element binding protein/ERK and PKC signaling pathways regulates positively autophagic activity. The in vivo results obtained with wild-type mice treated with methimazole and perchlorate or thyroxine were consistent with in vitro results. Next, in thyroid-specific autophagy knockout mice treated with methimazole and perchlorate (that is, mice were placed under a stressed condition where enhanced autophagy was required) for 2 months, lower follicle sizes and lower thyroglobulin contents in thyrocytes were observed, suggesting impaired thyroglobulin production presumably from insufficient nutrient supply. We therefore conclude that TSH positively regulates autophagic activity through the cAMP-PKA-cAMP response element binding protein/ERK and PKC signaling pathways, whereas thyroid hormones inhibit its activity in thyrocytes. Metabolites produced by autophagy appear to be necessary for protein synthesis stimulated by TSH.
Sigma-Aldrich
KT5720, A potent, specific, cell-permeable, reversible and ATP-competitive inhibitor of protein kinase A (Ki = 56 nM) that is prepared by a chemical modification of K-252a.
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)