Merck
  • p53 Is a Master Regulator of Proteostasis in SMARCB1-Deficient Malignant Rhabdoid Tumors.
p53 Is a Master Regulator of Proteostasis in SMARCB1-Deficient Malignant Rhabdoid Tumors.
Cancer cell (2019-02-13)
Alessandro Carugo, Rosalba Minelli, Luigi Sapio, Melinda Soeung, Federica Carbone, Frederick S Robinson, James Tepper, Ziheng Chen, Sara Lovisa, Maria Svelto, Samirkumar Amin, Sanjana Srinivasan, Edoardo Del Poggetto, Sara Loponte, Francesca Puca, Prasenjit Dey, Gabriel G Malouf, Xiaoping Su, Liren Li, Dolores Lopez-Terrada, Dinesh Rakheja, Alexander J Lazar, George J Netto, Priya Rao, Alessandro Sgambato, Anirban Maitra, Durga N Tripathi, Cheryl L Walker, Jose A Karam, Timothy P Heffernan, Andrea Viale, Charles W M Roberts, Pavlos Msaouel, Nizar M Tannir, Giulio F Draetta, Giannicola Genovese
ABSTRACT
Alterations in chromatin remodeling genes have been increasingly implicated in human oncogenesis. Specifically, the biallelic inactivation of the SWI/SNF subunit SMARCB1 results in the emergence of extremely aggressive pediatric malignancies. Here, we developed embryonic mosaic mouse models of malignant rhabdoid tumors (MRTs) that faithfully recapitulate the clinical-pathological features of the human disease. We demonstrated that SMARCB1-deficient malignancies exhibit dramatic activation of the unfolded protein response (UPR) and ER stress response via a genetically intact MYC-p19ARF-p53 axis. As a consequence, these tumors display an exquisite sensitivity to agents inducing proteotoxic stress and inhibition of the autophagic machinery. In conclusion, our findings provide a rationale for drug repositioning trials investigating combinations of agents targeting the UPR and autophagy in SMARCB1-deficient MRTs.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Venor GeM Mycoplasma Detection Kit, PCR-based
Sigma-Aldrich
Anti-SMARCB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution