Merck
  • Phenformin inhibits proliferation, invasion, and angiogenesis of cholangiocarcinoma cells via AMPK-mTOR and HIF-1A pathways.
Phenformin inhibits proliferation, invasion, and angiogenesis of cholangiocarcinoma cells via AMPK-mTOR and HIF-1A pathways.
Naunyn-Schmiedeberg's archives of pharmacology (2020-05-10)
Rattanaporn Jaidee, Sarinya Kongpetch, Laddawan Senggunprai, Auemduan Prawan, Upa Kukongviriyapan, Veerapol Kukongviriyapan
ABSTRACT
Phenformin (Phen), a potent activator of AMPK, is effective against some resistant cancers. This study evaluated the inhibition of proliferation, migration, invasion, and angiogenesis by Phen in aggressive cancer cells and investigated the underlying mechanism of the inhibition. Cholangiocarcinoma (CCA) KKU-156 and KKU-452 cells were used in this study. The results showed that Phen suppressed cell proliferation and induced apoptosis in both cells. Phen suppressed migration and invasion of cancer cells in wound healing and transwell chamber assays, respectively. The effects were associated with depletions of glutathione (GSH) and decreased glutathione redox ratio which represents cellular redox state. The redox stress was linked with the loss of mitochondrial transmembrane potential, as evaluated by JC-1 assay. The effect of Phen on angiogenesis was performed using HUVEC cultured cells. Phen alone did not affect tube formation of HUVEC cells. However, conditioned media from CCA cell cultures treated with Phen suppressed the tube-like structure formation. The antitumor effect of Phen was associated with AMPK activation and suppression of mTOR phosphorylation, HIF-1A, and VEGF protein expression. In conclusion, Phen inhibits cell proliferation, migration, invasion, and angiogenesis probably through AMPK-mTOR and HIF-1A-VEGF pathways. Phen may be repurposed as chemoprevention of cancer.
Sigma-Aldrich
Phenformin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Endothelial cell growth supplement from bovine neural tissue, ECGS, suitable for cell culture