Skip to Content
Beginning October 1st, 2024, the Life Science business of Merck Ltd. Korea will implement list price increases on our products. Learn More
Merck
  • Photobiomodulation therapy compensate the impairments of diabetic bone marrow mesenchymal stem cells.
Photobiomodulation therapy compensate the impairments of diabetic bone marrow mesenchymal stem cells.
Lasers in medical science (2019-07-25)
Fatemeh Zare, Mohammad Bayat, Abbas Aliaghaei, Abbas Piryaei
ABSTRACT
Pathophysiologic conditions associated with diabetes mellitus affect mesenchymal stem cells (MSCs), and this phenomenon may lead to some diabetic secondary complications. The present study was conducted to evaluate the impact of photobiomodulation (PBM) on rat diabetic MSC (DMSC) behavior in vitro. For the purpose of PBM, we used helium-neon laser with a wavelength of 632.8 nm at three different energy densities (0.5, 1, 2 J/cm2) and radiation periodicity of once, twice, and thrice. The survival, proliferation, and apoptosis in the normal MSCs (NMSCs), DMSCs, and diabetic MSCs, which were laser irradiated (DMSCs+L), were assessed using MTT assay, Ki67 immunofluorescence staining, and TUNEL assay, respectively. Our results demonstrated that DMSCs have significantly lower survival (P < 0.05) and proliferation rates (P < 0.001), and dramatically higher population doubling time (PDT, P < 0.001) and apoptosis rates (P < 0.001) as compared to NMSCs. Moreover, PBM with energy density of 1 J/cm2 and the periodicity of 1 or 2 times could improve diabetic MSC capabilities in the term of survival, proliferation, and apoptosis. Considering these findings, it is suggested that PBM could improve the ability of diabetic MSCs in vitro prior to transplantation or may rise their capabilities in their native niche in vivo.
Sigma-Aldrich
Alloxan monohydrate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.