Merck
  • The cystine/glutamate antiporter xCT is a key regulator of EphA2 S897 phosphorylation under glucose-limited conditions.
The cystine/glutamate antiporter xCT is a key regulator of EphA2 S897 phosphorylation under glucose-limited conditions.
Cellular signalling (2019-06-04)
Koji Teramoto, Hironori Katoh
ABSTRACT
EphA2, which belongs to the Eph family of receptor tyrosine kinases, is overexpressed in a variety of human cancers. Serine 897 (S897) phosphorylation of EphA2 is known to promote cancer cell migration and proliferation in a ligand-independent manner. In this study, we show that glucose deprivation induces S897 phosphorylation of EphA2 in glioblastoma cells. The phosphorylation requires the activity of the cystine/glutamate antiporter xCT and reactive oxygen species (ROS)-dependent ERK and RSK activation. Furthermore, depletion of EphA2 in glioblastoma cells leads to decreased cell viability under glucose starvation. Our results suggest a role of EphA2 in glioblastoma cell viability under glucose-limited conditions.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid