Merck
  • Synthesis and Fluorescence Properties of Structurally Characterized Heterobimetalic Cu(II)⁻Na(I) Bis(salamo)-Based Complex Bearing Square Planar, Square Pyramid and Triangular Prism Geometries of Metal Centers.
Synthesis and Fluorescence Properties of Structurally Characterized Heterobimetalic Cu(II)⁻Na(I) Bis(salamo)-Based Complex Bearing Square Planar, Square Pyramid and Triangular Prism Geometries of Metal Centers.
Molecules (Basel, Switzerland) (2018-04-26)
Xiu-Yan Dong, Qing Zhao, Zhi-Li Wei, Hao-Ran Mu, Han Zhang, Wen-Kui Dong
ABSTRACT
A novel heterotrinuclear complex [Cu₂(L)Na(µ-NO₃)]∙CH₃OH∙CHCl₃ derived from a symmetric bis(salamo)-type tetraoxime H₄L having a naphthalenediol unit, was prepared and structurally characterized via means of elemental analyses, UV-Vis, FT-IR, fluorescent spectra and single-crystal X-ray diffraction. The heterobimetallic Cu(II)⁻Na(I) complex was acquired via the reaction of H₄L with 2 equivalents of Cu(NO₃)₂·2H₂O and 1 equivalent of NaOAc. Clearly, the heterotrinuclear Cu(II)⁻Na(I) complex has a 1:2:1 ligand-to-metal (Cu(II) and Na(I)) ratio. X-ray diffraction results exhibited the different geometric behaviors of the Na(I) and Cu(II) atoms in the heterotrinuclear complex; the both Cu(II) atoms are sited in the N₂O₂ coordination environments of fully deprotonated (L)4− unit. One Cu(II) atom (Cu1) is five-coordinated and possesses a geometry of slightly distorted square pyramid, while another Cu(II) atom (Cu2) is four-coordination possessing a square planar coordination geometry. Moreover, the Na(I) atom is in the O₆ cavity and adopts seven-coordination with a geometry of slightly distorted single triangular prism. In addition, there are abundant supramolecular interactions in the Cu(II)⁻Na(I) complex. The fluorescence spectra showed the Cu(II)⁻Na(I) complex possesses a significant fluorescent quenching and exhibited a hypsochromic-shift compared with the ligand H₄L.
Sigma-Aldrich
o-Vanillin, 99%
Sigma-Aldrich
Boron tribromide, ReagentPlus®, 99.9%
Sigma-Aldrich
Methyltrioctylammonium chloride, ≥97.0% (AT)
Sigma-Aldrich
Pyridinium chlorochromate, 98%