
Terbufos-sulfoxide

Empirical Formula (Hill Notation):
C9H21O3PS3
CAS Number:
Molecular Weight:
304.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2