Skip to Content
Merck
(S,S)-(−)-Hydrobenzoin 99%, optical purity ee: 99% (GLC)

(S,S)-(−)-Hydrobenzoin

(S,S)-(−)-Hydrobenzoin 99%, optical purity ee: 99% (GLC)
Synonym(s):
(S,S)-(−)-1,2-Diphenyl-1,2-ethanediol, (S,S)-1,2-Diphenylethylene glycol
Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing