
Spermidine phosphate salt hexahydrate

Synonym(s):
Dispermidine triphosphate
Linear Formula:
[NH2(CH2)3NH(CH2)4NH2]2 · 3H3PO4 · 6H2O
CAS Number:
Molecular Weight:
346.28
Products
Product Category
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing