
Propyl Gallate Impurity A

Synonym(s):
Gallic acid, 3,4,5-Trihydroxybenzoic acid
Linear Formula:
(HO)3C6H2CO2H
CAS Number:
Molecular Weight:
170.12
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing