
Poly(methyl methacrylate-co-ethylene glycol dimethacrylate)

Linear Formula:
[-CH2C(CH3)(CO2CH3)-]X[CH2(C-)(CH3)CO2CH2CH2O2C(C-)(CH3)CH2-]Y
CAS Number:
Products
Physical Form
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing