
Poly(D,L-lactide-co-glycolide) acid terminated

Empirical Formula (Hill Notation):
(C3H4O2)x(C2H2O2)y
CAS Number:
Products
Brand
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing