
Paraoxon-methyl

Synonym(s):
O,O-Dimethyl O-(4-nitrophenyl) phosphate
Empirical Formula (Hill Notation):
C8H10NO6P
CAS Number:
Molecular Weight:
247.14
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2