
Oxymorphone

Synonym(s):
4,5α-Epoxy-3,14-dihydroxy-17-methylmorphinan-6-one, Dihydroxymorphinone
Empirical Formula (Hill Notation):
C17H19NO4
CAS Number:
Molecular Weight:
301.34
Products
Brand
Formula Weight
Manufacturer
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing