Skip to Content
Merck
Oxycodone Related Compound B pharmaceutical secondary standard

Oxycodone Related Compound B

Oxycodone Related Compound B pharmaceutical secondary standard
Synonym(s):
Oxycodone N-oxide, (5α)-4,5-Epoxy-14-hydroxy-3-methoxy-17-methylmorphinan-6-one 17-oxide
Empirical Formula (Hill Notation):
C18H21NO5
CAS Number:
Molecular Weight:
331.36

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing