
Oleic acid-1-13C

Linear Formula:
CH3(CH2)7CH=CH(CH2)713CO2H
CAS Number:
Molecular Weight:
283.45
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing