
N-Benzyl-L-proline ethyl ester

Empirical Formula (Hill Notation):
C14H19NO2
CAS Number:
Molecular Weight:
233.31
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing