
Monoclonal Anti-Interleukin-2 antibody produced in mouse

Synonym(s):
Anti-IL2, Anti-Lymphokine, Anti-TCGF
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing