Skip to Content
DO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mandarin oil
CAS Number:
8008-31-9
Products
Product Category
essential oils (2)
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Boiling Point (°C)
Color
Feature
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Kosher (2)
Natural (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W265722
Mandarin oil,
natural, distilled, product of Simone Gatto
Expand
View Pricing
W265799
Mandarin oil,
colorless, natural, product of Simone Gatto
Expand
View Pricing