
Levodopa Related Compound B

Synonym(s):
DL-3-O-Methyldopa, 3-Methoxytyrosine
Empirical Formula (Hill Notation):
C10H13NO4
CAS Number:
Molecular Weight:
211.21
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2