Skip to Content
Merck
Guaifenesin European Pharmacopoeia (EP) Reference Standard

Guaifenesin

Guaifenesin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Guaiacol glyceryl ether, 3-(o-Methoxyphenoxy)-1,2-propanediol, Glycerol guaiacolate, Guaifenesin
Empirical Formula (Hill Notation):
C10H14O4
CAS Number:
Molecular Weight:
198.22

Products

Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique