
Gadobutrol monohydrate

Empirical Formula (Hill Notation):
C18H31GdN4O9 · H2O
CAS Number:
Molecular Weight:
622.73
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing