Skip to Content
Merck
Fluroxypyr PESTANAL®, analytical standard

Fluroxypyr

Fluroxypyr PESTANAL®, analytical standard
Synonym(s):
2-[(4-Amino-3,5-dichloro-6-fluoro-2-pyridinyl)oxy]acetic acid, 4-Amino-3,5-dichloro-6-fluoro-2-pyridyloxyacetic acid
Empirical Formula (Hill Notation):
C7H5Cl2FN2O3
CAS Number:
Molecular Weight:
255.03

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing