Skip to Content
Merck
Ethanol-150 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®

Ethanol-150

Ethanol-150 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Synonym(s):
Ethanol solution, Ethyl alcohol
Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1