Skip to Content
Merck
D-Valine BioReagent, suitable for cell culture

D-Valine

D-Valine BioReagent, suitable for cell culture
Synonym(s):
(R)-α-Aminoisovaleric acid, D-2-Amino-3-methylbutanoic acid
Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing